Learn More
Tetra-n-butylammonium trifluoromethanesulfonate, 99%
CAS: 35895-70-6 | C18H36F3NO2 | 355.49 g/mol
Supplier: Thermo Scientific Chemicals A1101314
Description
Used as a catalyst for the reactions of condensation of alcohols and carboxylic acids, reaction of aromatic compounds with sulfonyl chlorides, cracking of alkanes, alkylation of alkenes, isomerisation of alkanes and trans-alkylation of aromatics, trans-bromination and other Friedel-Crafts reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as a catalyst for the reactions of condensation of alcohols and carboxylic acids, reaction of aromatic compounds with sulfonyl chlorides, cracking of alkanes, alkylation of alkenes, isomerisation of alkanes and trans-alkylation of aromatics, trans-bromination and other Friedel-Crafts reactions.
Solubility
Miscible in watetr. Soluble in methanol(0.1 g/mL).
Notes
Hygroscopic. Keep container tightly closed in a cool, dry and well-ventilated place. Store away from oxidizing agents, air and water.
Specifications
| Tetra-n-butylammonium trifluoromethanesulfonate | |
| 112°C to 115°C | |
| MFCD00042585 | |
| 4172978 | |
| tetrabutylammonium triflate, tetrabutylammonium trifluoromethanesulfonate, tetra-n-butylammonium trifluoromethanesulfonate, tetrabutylammonium ion triflate, trifluoromethanesulfonic acid tetrabutylammonium salt, acmc-1aglb, tetrakis but-1-yl ammonium triflate, tetrabutylazanium; trifluoromethanesulfonate, tetra-n-butylammonium trifluoromethanesulphonate | |
| WTEXQPWIUJQYJQ-UHFFFAOYSA-M | |
| tetrabutylazanium;trifluoromethanesulfonate | |
| 2733306 | |
| 99% |
| 35895-70-6 | |
| C18H36F3NO2 | |
| 25 g | |
| Hygroscopic | |
| Miscible in watetr. Soluble in methanol(0.1g/mL). | |
| [O-]C(=O)C(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC | |
| 355.49 | |
| 391.54 |
Chemical Identifiers
| 35895-70-6 | |
| 355.49 | |
| WTEXQPWIUJQYJQ-UHFFFAOYSA-M | |
| 2733306 | |
| [O-]C(=O)C(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC |
| C18H36F3NO2 | |
| MFCD00042585 | |
| tetrabutylammonium triflate, tetrabutylammonium trifluoromethanesulfonate, tetra-n-butylammonium trifluoromethanesulfonate, tetrabutylammonium ion triflate, trifluoromethanesulfonic acid tetrabutylammonium salt, acmc-1aglb, tetrakis but-1-yl ammonium triflate, tetrabutylazanium; trifluoromethanesulfonate, tetra-n-butylammonium trifluoromethanesulphonate | |
| tetrabutylazanium;trifluoromethanesulfonate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 252-783-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only