missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetra-n-butylammonium trifluoromethanesulfonate, 99%
Description
Used as a catalyst for the reactions of condensation of alcohols and carboxylic acids, reaction of aromatic compounds with sulfonyl chlorides, cracking of alkanes, alkylation of alkenes, isomerisation of alkanes and trans-alkylation of aromatics, trans-bromination and other Friedel-Crafts reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as a catalyst for the reactions of condensation of alcohols and carboxylic acids, reaction of aromatic compounds with sulfonyl chlorides, cracking of alkanes, alkylation of alkenes, isomerisation of alkanes and trans-alkylation of aromatics, trans-bromination and other Friedel-Crafts reactions.
Solubility
Miscible in watetr. Soluble in methanol(0.1 g/mL).
Notes
Hygroscopic. Keep container tightly closed in a cool, dry and well-ventilated place. Store away from oxidizing agents, air and water.
Used as a catalyst for the reactions of condensation of alcohols and carboxylic acids, reaction of aromatic compounds with sulfonyl chlorides, cracking of alkanes, alkylation of alkenes, isomerisation of alkanes and trans-alkylation of aromatics, trans-bromination and other Friedel-Crafts reactions.
Solubility
Miscible in watetr. Soluble in methanol(0.1 g/mL).
Notes
Hygroscopic. Keep container tightly closed in a cool, dry and well-ventilated place. Store away from oxidizing agents, air and water.
Specifications
Specifications
| Melting Point | 112°C to 115°C |
| Quantity | 5 g |
| Beilstein | 4172978 |
| Sensitivity | Hygroscopic |
| Solubility Information | Miscible in watetr. Soluble in methanol(0.1g/mL). |
| Formula Weight | 391.54 |
| Percent Purity | 99% |
| Chemical Name or Material | Tetra-n-butylammonium trifluoromethanesulfonate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 252-783-5 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only