missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium tribromide, 98+%

CAS: 38932-80-8 | C16H36Br3N | 482.15 g/mol

$348.69 - $348.69

Chemical Identifiers

CAS 38932-80-8
Molecular Formula C16H36Br3N
Molecular Weight (g/mol) 482.15
InChI Key SFLXUZPXEWWQNH-UHFFFAOYSA-K
Synonym tetrabutylammonium tribromide, tetra-n-butylammonium tribromide, tetrabutylammoniumtribromide, mono tetrabutylammonium tribromide, tetrabutylamoniumtribromide, acmc-1ct1n, tetrabutylamonium tribromide, tetrabutylazanium tribromide, tetrabutylammonium-tribromide, tetra-butylammonium tribromide
PubChem CID 23500184
IUPAC Name tetrabutylazanium;tribromide
SMILES CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.[Br-].[Br-].[Br-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC123971000
SDS View Documents
Thermo Scientific Chemicals
123971000
100 g Glass bottle
Each for $348.69
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Chemical Identifiers

38932-80-8
482.15
tetrabutylammonium tribromide, tetra-n-butylammonium tribromide, tetrabutylammoniumtribromide, mono tetrabutylammonium tribromide, tetrabutylamoniumtribromide, acmc-1ct1n, tetrabutylamonium tribromide, tetrabutylazanium tribromide, tetrabutylammonium-tribromide, tetra-butylammonium tribromide
tetrabutylazanium;tribromide
C16H36Br3N
SFLXUZPXEWWQNH-UHFFFAOYSA-K
23500184
CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.[Br-].[Br-].[Br-]

Specifications

38932-80-8
Orange
98% min. (Iodometry)
C16H36Br3N
100 g
Solubility in water: insoluble in water. Other solubilities: slightly soluble in alcohols, well soluble in acetonitrille,chcl3,thf,dioxane
CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC.[Br-].[Br-].[Br-]
482.15
482.15
Crystalline Powder
71.0°C to 75.0°C
Authentic
Glass bottle
[CH3(CH2)3]4NBr3
tetrabutylammonium tribromide, tetra-n-butylammonium tribromide, tetrabutylammoniumtribromide, mono tetrabutylammonium tribromide, tetrabutylamoniumtribromide, acmc-1ct1n, tetrabutylamonium tribromide, tetrabutylazanium tribromide, tetrabutylammonium-tribromide, tetra-butylammonium tribromide
SFLXUZPXEWWQNH-UHFFFAOYSA-K
tetrabutylazanium;tribromide
23500184
98+%
Tetra-n-butylammonium tribromide
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only