missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium tetrafluoroborate, 99%

Catalog No. AAA1668806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1668806 5 g
AAA1668814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1668806 Supplier Thermo Scientific Chemicals Supplier No. A1668806

CAS: 429-42-5 | C16H36BF4N | 329.27 g/mol

As Pharmaceutical Intermediates. Also in catalysis. Reasonably effiicient (1500-2000 plates/m), long-llved open tubular columns were prepared by using the organic molten salt tetra-n-butylammonlum tetrafluoroborate as the stationary phase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
As Pharmaceutical Intermediates. Also in catalysis. Reasonably effiicient (1500-2000 plates/m), long-llved open tubular columns were prepared by using the organic molten salt tetra-n-butylammonlum tetrafluoroborate as the stationary phase.

Solubility
Slightly soluble in Water, methanol( 0.1 g/mL), ethanol (100 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 429-42-5
Molecular Formula C16H36BF4N
Molecular Weight (g/mol) 329.27
MDL Number MFCD00011634
InChI Key NNZZSJSQYOFZAM-UHFFFAOYSA-N
Synonym tetrabutylammonium tetrafluoroborate, tetra-n-butylammonium tetrafluoroborate, tetrabutylammonium fluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-, tetrabutyl ammonium tetrafluoroborate, tetrabutylammonium ion tetrafluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-1:1, tbabf4, acmc-209jry, tbabf4;
PubChem CID 67932
IUPAC Name tetrabutylazanium;tetrafluoroborate
SMILES F[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 159°C to 162°C
Odor Odorless
Linear Formula [CH3(CH2)3]4NBF4
Quantity 5 g
Beilstein 3577508
Solubility Information Slightly soluble in Water,methanol( 0.1g/mL),ethanol (100mg/ml).
Formula Weight 329.27
Percent Purity 99%
Chemical Name or Material Tetra-n-butylammonium tetrafluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-058-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only