Learn More
Tetra-n-butylammonium sulfate, 50% w/w aq. soln.
Phase transfer catalyst, neutral pH | CAS: 2472-88-0 | C32H72N2O4S | 581.00 g/mol
$103.27 - $988.58
Chemical Identifiers
| CAS | 2472-88-0 |
|---|---|
| Molecular Formula | C32H72N2O4S |
| Molecular Weight (g/mol) | 581.00 |
| MDL Number | MFCD00216632 |
| InChI Key | ZXUCBXRTRRIBSO-UHFFFAOYSA-L |
| Synonym | bis tetrabutylammonium sulphate, tetrabutylammonium sulfate, tetrabutylammonium sulfate solution, 1-butanaminium, n,n,n-tributyl-, sulfate 2:1, tetrabutyl ammoniumsulfat, tetrabutylammonium sulphate, acmc-1cdo3, tetrabutyl ammonium sulfate, tetrabutyl-ammonium sulfate, tetra-n-butylammonium sulfate |
| PubChem CID | 11433187 |
| IUPAC Name | tetrabutylazanium;sulfate |
| SMILES | [O-]S([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA41723AE
|
Thermo Scientific Chemicals
041723AE |
100 mL |
Each for $103.27
|
|
|||||
|
AA41723AP
|
Thermo Scientific Chemicals
041723AP |
500 mL |
Each for $298.34
|
|
|||||
|
AA41723K4
|
Thermo Scientific Chemicals
041723K4 |
2.5 L |
Each for $988.58
|
|
|||||
Description
Tetra-n-butylammonium sulfate is used as a phase transfer catalyst, neutral pH and organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 2472-88-0 | |
| 581.00 | |
| ZXUCBXRTRRIBSO-UHFFFAOYSA-L | |
| 11433187 | |
| [O-]S([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC |
| C32H72N2O4S | |
| MFCD00216632 | |
| bis tetrabutylammonium sulphate, tetrabutylammonium sulfate, tetrabutylammonium sulfate solution, 1-butanaminium, n,n,n-tributyl-, sulfate 2:1, tetrabutyl ammoniumsulfat, tetrabutylammonium sulphate, acmc-1cdo3, tetrabutyl ammonium sulfate, tetrabutyl-ammonium sulfate, tetra-n-butylammonium sulfate | |
| tetrabutylazanium;sulfate |
Specifications
| 2472-88-0 | |
| 84°C | |
| C32H72N2O4S | |
| MFCD00216632 | |
| bis tetrabutylammonium sulphate, tetrabutylammonium sulfate, tetrabutylammonium sulfate solution, 1-butanaminium, n,n,n-tributyl-, sulfate 2:1, tetrabutyl ammoniumsulfat, tetrabutylammonium sulphate, acmc-1cdo3, tetrabutyl ammonium sulfate, tetrabutyl-ammonium sulfate, tetra-n-butylammonium sulfate | |
| ZXUCBXRTRRIBSO-UHFFFAOYSA-L | |
| tetrabutylazanium;sulfate | |
| 11433187 | |
| 50% w/w aq. soln. | |
| Tetra-n-butylammonium sulfate |
| 1.01 | |
| Unpleasant | |
| 1.417 | |
| 100 mL | |
| Fully miscible. | |
| [O-]S([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC | |
| 581.00 | |
| 580.99 | |
| Liquid |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 219-593-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only