missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium sulfate, 50% w/w aq. soln.

Phase transfer catalyst, neutral pH | CAS: 2472-88-0 | C32H72N2O4S | 581.00 g/mol

Supplier:  Thermo Scientific Chemicals 041723AP

 View more versions of this product

Catalog No. AA41723AP


Only null left
Explore available promotions

Description

Description

Tetra-n-butylammonium sulfate is used as a phase transfer catalyst, neutral pH and organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tetra-n-butylammonium sulfate
1.01
Unpleasant
1.417
500 mL
Fully miscible.
[O-]S([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC
581.00
580.99
Liquid
2472-88-0
84°C
C32H72N2O4S
MFCD00216632
bis tetrabutylammonium sulphate, tetrabutylammonium sulfate, tetrabutylammonium sulfate solution, 1-butanaminium, n,n,n-tributyl-, sulfate 2:1, tetrabutyl ammoniumsulfat, tetrabutylammonium sulphate, acmc-1cdo3, tetrabutyl ammonium sulfate, tetrabutyl-ammonium sulfate, tetra-n-butylammonium sulfate
ZXUCBXRTRRIBSO-UHFFFAOYSA-L
tetrabutylazanium;sulfate
11433187
50% w/w aq. soln.

Chemical Identifiers

2472-88-0
581.00
ZXUCBXRTRRIBSO-UHFFFAOYSA-L
11433187
[O-]S([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC.CCCC[N+](CCCC)(CCCC)CCCC
C32H72N2O4S
MFCD00216632
bis tetrabutylammonium sulphate, tetrabutylammonium sulfate, tetrabutylammonium sulfate solution, 1-butanaminium, n,n,n-tributyl-, sulfate 2:1, tetrabutyl ammoniumsulfat, tetrabutylammonium sulphate, acmc-1cdo3, tetrabutyl ammonium sulfate, tetrabutyl-ammonium sulfate, tetra-n-butylammonium sulfate
tetrabutylazanium;sulfate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 219-593-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only