Learn More
Tetra-n-butylammonium nitrate, 98%
CAS: 1941-27-1 | C16H36N2O3 | 304.475 g/mol
Supplier: Thermo Scientific Chemicals 04334214
Description
Tetra-n-butylammonium nitrate is used in coatings, printing ink, rubber, glass, leather and cosmetics. It serves as a supporting electrolyte in liquid ammonia. It is also used in the preparation of nitrates from triflates. It is employed in the inversion of configuration of alcohols through tosylates. It is also a source of NO3- used in nucleophilic substitution reactions. Further, it is used in the conversion of halides into ketones. In addition to this, it is used as a source of nitronium ions for the nitration of olefins, amides, ribonucleosides, glycols and aromatic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Tetra-n-butylammonium nitrate | |
| 116°C to 118°C | |
| C16H36N2O3 | |
| 25 g | |
| Hygroscopic | |
| Soluble in dichloromethane,acetonitrile,benzene and toluene. | |
| CCCC[N+](CCCC)(CCCC)CCCC.[N+](=O)([O-])[O-] | |
| 304.475 | |
| 304.48 | |
| Powder |
| 1941-27-1 | |
| Weak | |
| MFCD00043202 | |
| UN1479 | |
| tetrabutylammonium nitrate, tetra-n-butylammonium nitrate, 1-butanaminium, n,n,n-tributyl-, nitrate, tetrabutylazanium nitrate, ammonium, tetrabutyl-, nitrate, n-n-n-tributyl-1-butanaminium nitrate, 1-butanaminium, n,n,n-tributyl-, nitrate 1:1, tetrabutylammonium ion nitrate, tetrabutylammoniumnitrate, acmc-209exx | |
| QHOKENWFMZXSEU-UHFFFAOYSA-N | |
| tetrabutylazanium;nitrate | |
| 16027 | |
| 98% |
Chemical Identifiers
| 1941-27-1 | |
| 304.475 | |
| QHOKENWFMZXSEU-UHFFFAOYSA-N | |
| 16027 | |
| CCCC[N+](CCCC)(CCCC)CCCC.[N+](=O)([O-])[O-] |
| C16H36N2O3 | |
| MFCD00043202 | |
| tetrabutylammonium nitrate, tetra-n-butylammonium nitrate, 1-butanaminium, n,n,n-tributyl-, nitrate, tetrabutylazanium nitrate, ammonium, tetrabutyl-, nitrate, n-n-n-tributyl-1-butanaminium nitrate, 1-butanaminium, n,n,n-tributyl-, nitrate 1:1, tetrabutylammonium ion nitrate, tetrabutylammoniumnitrate, acmc-209exx | |
| tetrabutylazanium;nitrate |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P220-P221-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P372-P375-P380-P501c
H272-H315-H319-H335
DOTInformation : Hazard Class: 5.1; Packaging Group: II
EINECSNumber : 217-726-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only