missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetra-n-butylammonium hydroxide, 40% w/w aq. soln.
Description
Tetra-n-butylammonium hydroxide, 40% w/w aq. solution is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is used as a curing accelerator for epoxy resins. It is also used in preparation of dichlorocarbene from chloroform.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetra-n-butylammonium hydroxide, 40% w/w aq. solution is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is used as a curing accelerator for epoxy resins. It is also used in preparation of dichlorocarbene from chloroform.
Solubility
Miscible with organic solvents.
Notes
Air sensitive and hygroscopic. Incompatible with strong acids and corrodes metal.
Tetra-n-butylammonium hydroxide, 40% w/w aq. solution is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is used as a curing accelerator for epoxy resins. It is also used in preparation of dichlorocarbene from chloroform.
Solubility
Miscible with organic solvents.
Notes
Air sensitive and hygroscopic. Incompatible with strong acids and corrodes metal.
Specifications
Specifications
| Density | 0.99 |
| pH | 14.0 |
| Flash Point | None |
| Odor | Amine-like |
| Refractive Index | 1.408 |
| Linear Formula | [CH3(CH2)3]4NOH |
| Quantity | 50 g |
| UN Number | UN3267 |
| Beilstein | 3571228 |
| Sensitivity | Air Sensitive |
| Show More |
Safety and Handling
| Hazard Category | H302-H314-H335 |
| Hazard Statement | GHS H Statement H314 Causes severe skin burns and eye damage. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S. |
| EINECSNumber | 218-147-6 |
| RTECSNumber | BS5425000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only