missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium hydroxide, 40% w/w aq. soln.

Catalog No. AAL0280918
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0280918 50 g
AAL0280930 250 g
AAL028090B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0280918 Supplier Thermo Scientific Chemicals Supplier No. L0280918
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2052-49-5 | C16H37NO | 259.48 g/mol

Tetra-n-butylammonium hydroxide, 40% w/w aq. solution is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is used as a curing accelerator for epoxy resins. It is also used in preparation of dichlorocarbene from chloroform.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-butylammonium hydroxide, 40% w/w aq. solution is used as a phase-transfer catalyst in organic synthesis especially for alkylation and condensation reactions. It is used as a curing accelerator for epoxy resins. It is also used in preparation of dichlorocarbene from chloroform.

Solubility
Miscible with organic solvents.

Notes
Air sensitive and hygroscopic. Incompatible with strong acids and corrodes metal.

Chemical Identifiers

CAS 2052-49-5
Molecular Formula C16H37NO
Molecular Weight (g/mol) 259.48
MDL Number MFCD00009425
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
Synonym tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
PubChem CID 2723671
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Density 0.99
pH 14.0
Flash Point None
Odor Amine-like
Refractive Index 1.408
Linear Formula [CH3(CH2)3]4NOH
Quantity 50 g
UN Number UN3267
Beilstein 3571228
Sensitivity Air Sensitive
Solubility Information Miscible with organic solvents.
IUPAC Name tetrabutylazanium hydroxide
Formula Weight 259.48
Concentration or Composition (by Analyte or Components) 40% w/w aq. soln.
Chemical Name or Material Tetra-n-butylammonium hydroxide
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
EINECSNumber 218-147-6
RTECSNumber BS5425000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only