missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium hydroxide, 1.0M aq. soln., HPLC grade

Catalog No. p-7058955
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43662AC 25 mL
AA43662AE 100 mL
AA43662AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA43662AC Supplier Thermo Scientific Chemicals Supplier No. 043662AC
Only null left

Phase transfer catalyst, electrolyte in polarography, ion-pair extraction | CAS: 2052-49-5 | C16H37NO | 259.48 g/mol

Phase transfer catalyst, electrolyte in polarography, ion-pair extractionTetra-n-butylammonium hydroxide, 1.0M in aqueous solution is used as a strong base in organic chemistry. It is utilized under phase-transfer conditions to effect alkylation and deprotonation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2052-49-5
Molecular Formula C16H37NO
Molecular Weight (g/mol) 259.48
MDL Number MFCD00009425
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
Synonym tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
PubChem CID 2723671
IUPAC Name tetrabutylazanium;hydroxide
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Flash Point None
Linear Formula [CH3(CH2)3]4NOH
Quantity 25 mL
UN Number UN3267
Beilstein 3571228
Sensitivity Air Sensitive
Solubility Information Miscible with organic solvents.
Formula Weight 259.48
Grade HPLC
Concentration or Composition (by Analyte or Components) 1.0M aq. soln.
Physical Form Liquid
Chemical Name or Material Tetra-n-butylammonium hydroxide
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314
Causes severe skin burns and eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
EINECSNumber 218-147-6
RTECSNumber BS5425000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only