missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium hydrogen sulfate, 97%

Catalog No. AAA1404718
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1404718 50 g
AAA1404730 250 g
AAA140470B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1404718 Supplier Thermo Scientific Chemicals Supplier No. A1404718
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 32503-27-8 | C16H37NO4S | 339.54 g/mol

Phase transfer catalyst Tetra-n-butylammonium hydrogen sulfate shows antibacterial properties due to the presence of the quaternary amine and the counter ion. It serves as a phase-transfer catalyst, surface-active agent, solvent, intermediate, active ingredient for conditioners, antistatic agent, detergent sanitizers, softener for textiles and paper products, emulsifying agents and pigment dispersers. It is used as an efficient catalyst in the N-alkylation reactions of benzanilides, cyclization of beta-amino acids to beta-lactams, conversion of nitriles to amides, dehydrohalogenation of aryl 2-haloethyl ethers to give vinyl ethers and in the oxidation of alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phase transfer catalyst Tetra-n-butylammonium hydrogen sulfate shows antibacterial properties due to the presence of the quaternary amine and the counter ion. It serves as a phase-transfer catalyst, surface-active agent, solvent, intermediate, active ingredient for conditioners, antistatic agent, detergent sanitizers, softener for textiles and paper products, emulsifying agents and pigment dispersers. It is used as an efficient catalyst in the N-alkylation reactions of benzanilides, cyclization of beta-amino acids to beta-lactams, conversion of nitriles to amides, dehydrohalogenation of aryl 2-haloethyl ethers to give vinyl ethers and in the oxidation of alcohols.

Solubility
Soluble in water.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 32503-27-8
Molecular Formula C16H37NO4S
Molecular Weight (g/mol) 339.54
MDL Number MFCD00011637
InChI Key SHFJWMWCIHQNCP-UHFFFAOYSA-M
Synonym tetrabutylammonium hydrogen sulfate, tetrabutylammonium hydrogensulfate, tetrabutylammonium bisulfate, tetra-n-butylammonium hydrogen sulfate, tbahs, tetrabutylammonium sulfate 1:1, 1-butanaminium, n,n,n-tributyl-, sulfate 1:1, tetrabutylammonium hydrogen sulphate, tetrabutylazanium hydrogen sulfate, ipc-tba-hs
PubChem CID 94433
IUPAC Name hydrogen sulfate;tetrabutylazanium
SMILES OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 168°C to 172°C
Quantity 50 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 339.54
Percent Purity 97%
Chemical Name or Material Tetra-n-butylammonium hydrogen sulfate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 251-068-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only