missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium fluoride trihydrate, 98%

Catalog No. AAL1330309
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1330309 10 g
AAL1330318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1330309 Supplier Thermo Scientific Chemicals Supplier No. L1330309
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 87749-50-6 | C16H42FNO3 | 315.514 g/mol

Tetra-n-butylammonium fluoride trihydrate is used as a phase transfer catalyst, as a mild base and as a source of fluoride ion in organic solvents. It is also used as a deprotecting agent to remove silyl ether protecting groups. It acts as a reactant for the preparation of deprotecting agents, in preparation of cellulose derivatives, lipophilic peptides for DNA transfections and Dehydrobromination reactions. It plays an important role in the convertion of O-silylated enolates in to carbonyls in the presence of dimethyl sulfoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-butylammonium fluoride trihydrate is used as a phase transfer catalyst, as a mild base and as a source of fluoride ion in organic solvents. It is also used as a deprotecting agent to remove silyl ether protecting groups. It acts as a reactant for the preparation of deprotecting agents, in preparation of cellulose derivatives, lipophilic peptides for DNA transfections and Dehydrobromination reactions. It plays an important role in the convertion of O-silylated enolates in to carbonyls in the presence of dimethyl sulfoxide.

Solubility
Soluble in water, terahydrofuran, dimethyl sulfoxide and acetonitrile.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 87749-50-6
Molecular Formula C16H42FNO3
Molecular Weight (g/mol) 315.514
MDL Number MFCD00149981
InChI Key VEPTXBCIDSFGBF-UHFFFAOYSA-M
Synonym tetrabutylammonium fluoride trihydrate, tetra-n-butylammonium fluoride trihydrate, tetrabutylazanium fluoride trihydrate, tetrabutylammonium trihydrate fluoride, 1-butanaminium, n,n,n-tributyl-, fluoride, trihydrate, tetrabutylammonium ion trihydrate fluoride, tetra-n-butylammonium fluoride hydrate, bu4nf trihydrate, pubchem2275, pubchem4057
PubChem CID 11726816
IUPAC Name tetrabutylazanium;fluoride;trihydrate
SMILES CCCC[N+](CCCC)(CCCC)CCCC.O.O.O.[F-]

Specifications

Melting Point 54°C to 60°C
Odor Odorless
Linear Formula [CH3(CH2)3]4NF·3H2O
Quantity 10 g
UN Number UN2811
Beilstein 3761900
Sensitivity Hygroscopic
Solubility Information Soluble in water,terahydrofuran,dimethyl sulfoxide and acetonitrile.
Formula Weight 315.52 (261.47 Anhydrous)
Percent Purity 98%
Chemical Name or Material Tetra-n-butylammonium fluoride trihydrate
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H300-H318-H315
Fatal if swallowed.
Causes serious eye damage.
Causes skin irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 207-057-2
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only