missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butoxysilane, 97%

CAS: 4766-57-8 | C16H36O4Si | 320.545 g/mol

$76.63 - $213.60

Chemical Identifiers

CAS 4766-57-8
Molecular Formula C16H36O4Si
Molecular Weight (g/mol) 320.545
MDL Number MFCD00009432
InChI Key UQMOLLPKNHFRAC-UHFFFAOYSA-N
Synonym tetrabutyl orthosilicate, tetrabutoxysilane, tetra-n-butoxysilane, silane, tetrabutoxy, silicon tetrabutoxide, butyl silicate buo 4si, silicic acid h4sio4 , tetrabutyl ester, butyl silicate c4h9o 4si, butyl silicate, tetrabutyl orthosilice
PubChem CID 78500
IUPAC Name tetrabutyl silicate
SMILES CCCCO[Si](OCCCC)(OCCCC)OCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0314114
View Documents
Thermo Scientific Chemicals
L0314114
25 g
Each for $76.63
Only null left
 
AAL0314122
View Documents
Thermo Scientific Chemicals
L0314122
100 g
Each for $213.60
Only null left
 
Description

Description

It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields and also in the alkylation and alkoxylation of silane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields and also in the alkylation and alkoxylation of silane.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications

Chemical Identifiers

4766-57-8
320.545
UQMOLLPKNHFRAC-UHFFFAOYSA-N
78500
CCCCO[Si](OCCCC)(OCCCC)OCCCC
C16H36O4Si
MFCD00009432
tetrabutyl orthosilicate, tetrabutoxysilane, tetra-n-butoxysilane, silane, tetrabutoxy, silicon tetrabutoxide, butyl silicate buo 4si, silicic acid h4sio4 , tetrabutyl ester, butyl silicate c4h9o 4si, butyl silicate, tetrabutyl orthosilice
tetrabutyl silicate

Specifications

277°C
4766-57-8
C16H36O4Si
1709274
tetrabutyl orthosilicate, tetrabutoxysilane, tetra-n-butoxysilane, silane, tetrabutoxy, silicon tetrabutoxide, butyl silicate buo 4si, silicic acid h4sio4 , tetrabutyl ester, butyl silicate c4h9o 4si, butyl silicate, tetrabutyl orthosilice
UQMOLLPKNHFRAC-UHFFFAOYSA-N
tetrabutyl silicate
78500
97%
1.413
0.899
<-80°C
25 g
MFCD00009432
135°C (275°F)
Not miscible or difficult to mix in water.
CCCCO[Si](OCCCC)(OCCCC)OCCCC
320.545
320.55
Characteristic
Moisture sensitive
Tetra-n-butoxysilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 225-305-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only