missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butoxysilane, 97%

CAS: 4766-57-8 | C16H36O4Si | 320.545 g/mol

Supplier:  Thermo Scientific Chemicals L0314122

 View more versions of this product

Catalog No. AAL0314122


Only null left
Explore available promotions

Description

Description

It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields and also in the alkylation and alkoxylation of silane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields and also in the alkylation and alkoxylation of silane.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture sensitive. Store away from oxidizing agents and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications

Specifications

277°C
<-80°C
100 g
MFCD00009432
135°C (275°F)
Not miscible or difficult to mix in water.
CCCCO[Si](OCCCC)(OCCCC)OCCCC
320.545
320.55
Characteristic
Moisture sensitive
Tetra-n-butoxysilane
4766-57-8
C16H36O4Si
1709274
tetrabutyl orthosilicate, tetrabutoxysilane, tetra-n-butoxysilane, silane, tetrabutoxy, silicon tetrabutoxide, butyl silicate buo 4si, silicic acid h4sio4 , tetrabutyl ester, butyl silicate c4h9o 4si, butyl silicate, tetrabutyl orthosilice
UQMOLLPKNHFRAC-UHFFFAOYSA-N
tetrabutyl silicate
78500
97%
1.413
0.899

Chemical Identifiers

4766-57-8
320.545
UQMOLLPKNHFRAC-UHFFFAOYSA-N
78500
CCCCO[Si](OCCCC)(OCCCC)OCCCC
C16H36O4Si
MFCD00009432
tetrabutyl orthosilicate, tetrabutoxysilane, tetra-n-butoxysilane, silane, tetrabutoxy, silicon tetrabutoxide, butyl silicate buo 4si, silicic acid h4sio4 , tetrabutyl ester, butyl silicate c4h9o 4si, butyl silicate, tetrabutyl orthosilice
tetrabutyl silicate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 225-305-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only