missing translation for 'onlineSavingsMsg'
Learn More

Tetra(decyl)ammonium Bromide 98.0+%, TCI America™

Catalog No. T114225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T114225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T114225G Supplier TCI America Supplier No. T114225G
Only null left

Chemical Identifiers

CAS 14937-42-9
Molecular Formula C40H84BrN
Molecular Weight (g/mol) 659.023
MDL Number MFCD00043166
InChI Key AHNISXOXSNAHBZ-UHFFFAOYSA-M
Synonym tetrakis decyl ammonium bromide, tetra decyl ammonium bromide, tetra-decylammonium bromide, tetra-n-decylammonium bromide, acmc-1c6fh, tetrakis-decylazanium bromide, tetrakis decyl azanium bromide, 1-decanaminium, n,n,n-tris decyl-, bromide
PubChem CID 3014876
IUPAC Name tetrakis-decylazanium;bromide
SMILES CCCCCCCCCC[N+](CCCCCCCCCC)(CCCCCCCCCC)CCCCCCCCCC.[Br-]

Specifications

Melting Point 88°C
Color White
Quantity 25 g
Formula Weight 659.02
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetra(decyl)ammonium Bromide