Learn More
Applied Biosystems™ TET™ Dye Phosphoramidite
Catalog No. p-7083029
Encompass
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
403171 50 mg
401533 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 403171 Supplier Applied Biosystems™ Supplier No. 403171
Only null left
Contains 100 mg TET™ Dye Phosphoramidite.TET™ Dye Phosphoramidite is used for the 5’ attachment of TET™ dye during oligonucleotide synthesis.The label is treated as if it were simply another base: It is inserted at the appropriate place in the sequence and incorporated automatically.
On-instrument labeling typically saves hours of post-synthesis manual processingFluorescent label TET™ is a single-isomer fluorescein dye designed for detection and analysis of labeled PCR products on ABI PRISM™ genetic analysis systems. TET™ is equally useful in applications historically employing isotopic labels. As such, it eliminates the hazard associated with handling, storage, and disposal of radioactive materials.
  • Specialty phosphoramidites allow online addition of the label during synthesis, saving time in production of labeled probes and primers
  • Designed and optimized for use with ABI™ synthesizers, guaranteeing reliable nucleic acid synthesis
  • We offer a range of specialty phosphoramites and linkers to give you the flexibility demanded in oligonucleotide synthesis
Specifications
Packaging Type Shipped on Dry Ice
Quantity 50 mg
For Research Use Only. Not for use in diagnostic procedures.