missing translation for 'onlineSavingsMsg'
Learn More

Testosterone Solution, 1.0 mg/mL in 1,2-dimethoxyethane, Drug Standard, MilliporeSigma™ Supelco™

Catalog No. 111017225
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111017225 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111017225 Supplier Merck Emd Millipore Supplier No. T54111ML

Specifications

Packaging Ampule
Concentration 1.0 mg/mL in 1,2-dimethoxyethane
MDL Number MFCD00003654
Quantity 1 mL
Grade Drug Standard