Learn More
Test mixture 2 for Apolar Capillary Columns (According to Grob for GC, ready-to-use), MilliporeSigma™ Supelco™
Mixture of D(-)-2,3-butanediol, decane, dicyclohexylamine, 2,6-dimethylaniline, 2,6-dimethylphenol, 2-ethylcaproic acid, methyl decanoate, methyl laurate, methyl undecanoate, 1-octanol and dodecane in hexane and methylene chloride
Supplier: Merck Emd Millipore 8650110ML
Specifications
| 110-54-3 | |
| MFCD00148603 | |
| 10 mL |
| -26°C - closed cup | |
| Analytical Standard | |
| Analytical Standard |