missing translation for 'onlineSavingsMsg'
Learn More

tert-Pentylmagnesium chloride, 1M in 2-MeTHF

Catalog No. AAH54824AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH54824AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH54824AE Supplier Thermo Scientific Chemicals Supplier No. H54824AE
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 28276-08-6 | C5H11ClMg | 130.90 g/mol

It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Store at 4°C. Air and moisture sensitive.

Chemical Identifiers

CAS 28276-08-6
Molecular Formula C5H11ClMg
Molecular Weight (g/mol) 130.90
MDL Number MFCD00010356
InChI Key NWFDYJDCSIHFAB-UHFFFAOYSA-M
Synonym 1,1-dimethylpropylmagnesium chloride, magnesium, chloro 1,1-dimethylpropyl, t-amylmagnesium chloride, tert-pentylmagnesium chloride, nwfdyjdcsihfab-uhfffaoysa-m, 1,1-dimethylpropyl magnesium chloride, magnesium,chloro 1,1-dimethylpropyl, chloro 2-methylbutan-2-yl magnesium, tert-pentylmagnesium chloride, 1m in 2-methf, 1,1-dimethylpropylmagnesium chloride solution, 1.0 m in diethyl ether
PubChem CID 4056867
IUPAC Name magnesium;2-methylbutane;chloride
SMILES CCC(C)(C)[Mg]Cl

Specifications

Density 0.831
Linear Formula CH3CH2C(CH3)2MgCl
Quantity 100 mL
UN Number UN3399
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 130.9
Concentration or Composition (by Analyte or Components) 1M in 2-MeTHF
Chemical Name or Material tert-Pentylmagnesium chloride
Hazard Category H225-H260-H302-H314-H335
Hazard Statement GHS H Statement
H225-H261-H314
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only