CAS RN 28276-08-6
IUPAC Name:
chloro(2-methylbutan-2-yl)magnesium
Synonyms:
chloro(2-methylbutan-2-yl)magnesium
Molecular Weight (g/mol):
130.9
Molecular Formula:
C5H11ClMg
InChi Key:
NWFDYJDCSIHFAB-UHFFFAOYSA-M
SMILES:
CCC(C)(C)[Mg]Cl
1
–
1
of
1
results
1
Promotions Available
tert-Pentylmagnesium chloride, 1M in 2-MeTHF
tert-Pentylmagnesium chloride, 1M in 2-MeTHF
CAS: 28276-08-6 Molecular Formula: C5H11ClMg Molecular Weight (g/mol): 130.90 MDL Number: MFCD00010356 InChI Key: NWFDYJDCSIHFAB-UHFFFAOYSA-M Synonym: 1,1-dimethylpropylmagnesium chloride,magnesium, chloro 1,1-dimethylpropyl,t-amylmagnesium chloride,tert-pentylmagnesium chloride,nwfdyjdcsihfab-uhfffaoysa-m,1,1-dimethylpropyl magnesium chloride,magnesium,chloro 1,1-dimethylpropyl,chloro 2-methylbutan-2-yl magnesium,tert-pentylmagnesium chloride, 1m in 2-methf,1,1-dimethylpropylmagnesium chloride solution, 1.0 m in diethyl ether PubChem CID: 4056867 IUPAC Name: magnesium;2-methylbutane;chloride SMILES: CCC(C)(C)[Mg]Cl