missing translation for 'onlineSavingsMsg'
Learn More

tert-Nitrobutane, 98+%

Catalog No. AAA1960822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1960822 100 g
AAA1960806 5 g
AAA1960814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1960822 Supplier Thermo Scientific Chemicals Supplier No. A1960822
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 594-70-7 | C4H9NO2 | 103.121 g/mol

It is used in the cyclic voltammetric study of tert-nitrobutane reduction in acetonitrile at mercury and platinum electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the cyclic voltammetric study of tert-nitrobutane reduction in acetonitrile at mercury and platinum electrodes.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 594-70-7
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00007393
InChI Key AIMREYQYBFBEGQ-UHFFFAOYSA-N
Synonym propane, 2-methyl-2-nitro, trimethylnitromethane, 2-nitroisobutane, 2-nitro-2-methylpropane, tert-nitrobutane, 1,1-dimethyl-1-nitroethane, nitro-tert-butane, 2-methyl-2-nitro-propane, unii-9dkh5l679z, ccris 5044
PubChem CID 11672
IUPAC Name 2-methyl-2-nitropropane
SMILES CC(C)(C)[N+](=O)[O-]

Specifications

Melting Point 27°C to 31°C
Density 0.95
Boiling Point 126°C to 127°C
Flash Point 19°C (66°F)
Refractive Index 1.4
Quantity 100 g
UN Number UN1325
Beilstein 1744184
Solubility Information Insoluble in water.
Formula Weight 103.12
Percent Purity ≥98%
Chemical Name or Material tert-Nitrobutane
Show More Show Less
Hazard Category H228-H315-H319-H335
Hazard Statement GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 209-851-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only