missing translation for 'onlineSavingsMsg'
Learn More

tert-Dodecyl Mercaptan (mixture of isomers) 98.0+%, TCI America™

Catalog No. D097125ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D097125ML 25 mL
D0971500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D097125ML Supplier TCI America Supplier No. D097125ML

Chemical Identifiers

CAS 25103-58-6
Molecular Formula C12H26S
MDL Number MFCD00043233
Synonym tert-Dodecanethiol, tert-Lauryl Mercaptan

Specifications

Color Yellow
Quantity 25 mL
UN Number 3082
Formula Weight 202.40
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material tert-Dodecyl Mercaptan (mixture of isomers)
EINECSNumber (2)-0464
RTECSNumber JR3150000
TSCA Yes