CAS RN 25103-58-6
IUPAC Name:
2,3,3,4,4,5-hexamethylhexane-2-thiol
Synonyms:
2,3,3,4,4,5-hexamethylhexane-2-thiol
Molecular Weight (g/mol):
202.4
Molecular Formula:
C12H26S
InChi Key:
YAJYJWXEWKRTPO-UHFFFAOYSA-N
SMILES:
CC(C)C(C)(C)C(C)(C)C(C)(C)S
1
–
1
of
1
results
1
tert-Dodecyl Mercaptan (mixture of isomers) 98.0+%, TCI America™
CAS: 25103-58-6 Molecular Formula: C12H26S MDL Number: MFCD00043233 Synonym: tert-Dodecanethiol, tert-Lauryl Mercaptan