missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylphosphonic acid, 98%

Catalog No. AC321520050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC321520050 5 g Glass bottle
AC321520010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC321520050 Supplier Thermo Scientific Chemicals Supplier No. 321520050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4923-84-6 | C4H9O3P | 136.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4923-84-6
Molecular Formula C4H9O3P
Molecular Weight (g/mol) 136.09
MDL Number MFCD00012296
InChI Key OGDSVONAYZTTDA-UHFFFAOYSA-L
Synonym phosphonic acid, 1,1-dimethylethyl, acmc-20ajka, t-butylphosphonic acid, tert-butylphosphonic acid, 2-methylprop-2-ylphosphonic acid, phosphonicacid, 1,1-dimethylethyl
PubChem CID 312546
IUPAC Name tert-butylphosphonic acid
SMILES CC(C)(C)P([O-])([O-])=O

Specifications

Melting Point 188.0°C to 192.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3)3CP(O)(OH)2
Quantity 5 g
Formula Weight 138.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material tert-Butylphosphonic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only