CAS RN 4923-84-6
IUPAC Name:
tert-butylphosphonate
Synonyms:
tert-butylphosphonate
Molecular Weight (g/mol):
136.09
Molecular Formula:
C4H9O3P
InChi Key:
OGDSVONAYZTTDA-UHFFFAOYSA-L
SMILES:
CC(C)(C)P([O-])([O-])=O
1
–
1
of
1
results
1
Promotions Available
tert-Butylphosphonic acid, 98%
tert-Butylphosphonic acid, 98%
CAS: 4923-84-6 Molecular Formula: C4H9O3P Molecular Weight (g/mol): 136.09 MDL Number: MFCD00012296 InChI Key: OGDSVONAYZTTDA-UHFFFAOYSA-L Synonym: phosphonic acid, 1,1-dimethylethyl,acmc-20ajka,t-butylphosphonic acid,tert-butylphosphonic acid,2-methylprop-2-ylphosphonic acid,phosphonicacid, 1,1-dimethylethyl PubChem CID: 312546 IUPAC Name: tert-butylphosphonic acid SMILES: CC(C)(C)P([O-])([O-])=O