Learn More
tert-Butylmagnesium chloride, 1M in MeTHF
CAS: 677-22-5 | C4H9ClMg | 116.87 g/mol
$330.11 - $330.11
Chemical Identifiers
| CAS | 677-22-5 |
|---|---|
| Molecular Formula | C4H9ClMg |
| Molecular Weight (g/mol) | 116.87 |
| MDL Number | MFCD00000465 |
| InChI Key | ZDRJSYVHDMFHSC-UHFFFAOYSA-M |
| Synonym | tert-butylmagnesium chloride, tert-butylmagnesiumchloride, t-butylmagnesium chloride, t-butyl magnesium chloride, tert-butyl chloro magnesium, magnesium, chloro 1,1-dimethylethyl, tert-butylmagnesium chloride solution, 1.0 m in thf, tbumgcl, tert-bumgcl, t-bumgcl |
| PubChem CID | 2724198 |
| IUPAC Name | magnesium;2-methylpropane;chloride |
| SMILES | CC(C)(C)[Mg]Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH51164AE
|
Thermo Scientific Chemicals
H51164AE |
100 mL |
Each for $330.11
|
|
|||||
Description
tert-Butylmagnesium chloride is a Grignard reagent used in organic synthesis. It is also involved in copper-catalyzed cross-coupling reaction with primary-alkyl halides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstert-Butylmagnesium chloride is a Grignard reagent used in organic synthesis. It is also involved in copper-catalyzed cross-coupling reaction with primary-alkyl halides.
Solubility
Miscible with alcohol and water.
Notes
Air and moisture sensitive. Incompatible with oxidizing agents and bases.
Chemical Identifiers
| 677-22-5 | |
| 116.87 | |
| ZDRJSYVHDMFHSC-UHFFFAOYSA-M | |
| 2724198 | |
| CC(C)(C)[Mg]Cl |
| C4H9ClMg | |
| MFCD00000465 | |
| tert-butylmagnesium chloride, tert-butylmagnesiumchloride, t-butylmagnesium chloride, t-butyl magnesium chloride, tert-butyl chloro magnesium, magnesium, chloro 1,1-dimethylethyl, tert-butylmagnesium chloride solution, 1.0 m in thf, tbumgcl, tert-bumgcl, t-bumgcl | |
| magnesium;2-methylpropane;chloride |
Specifications
| 677-22-5 | |
| (CH3)3CMgCl | |
| 100 mL | |
| Air and moisture sensitive | |
| Miscible with alcohol and water. | |
| CC(C)(C)[Mg]Cl | |
| 116.87 | |
| 116.87 | |
| tert-Butylmagnesium chloride |
| C4H9ClMg | |
| MFCD00000465 | |
| UN3399 | |
| tert-butylmagnesium chloride, tert-butylmagnesiumchloride, t-butylmagnesium chloride, t-butyl magnesium chloride, tert-butyl chloro magnesium, magnesium, chloro 1,1-dimethylethyl, tert-butylmagnesium chloride solution, 1.0 m in thf, tbumgcl, tert-bumgcl, t-bumgcl | |
| ZDRJSYVHDMFHSC-UHFFFAOYSA-M | |
| magnesium;2-methylpropane;chloride | |
| 2724198 | |
| 1M in MeTHF |
Safety and Handling
GHS H Statement
H225-H261-H314
Highly flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c
H225-H260-H302-H314-H335
DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only