missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldiphenylsilyl Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. p-7119831
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28981G 1 g
B28985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28981G Supplier TCI America Supplier No. B28981G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 92886-86-7
Molecular Formula C17H19F3O3SSi
Molecular Weight (g/mol) 388.48
MDL Number MFCD01941212
InChI Key AOMOJFWVOHXBTN-UHFFFAOYSA-N
Synonym tert-Butyldiphenylsilyl Triflate, Trifluoromethanesulfonic Acid tert-Butyldiphenylsilyl Ester
PubChem CID 10949115
IUPAC Name tert-butyldiphenylsilyl trifluoromethanesulfonate
SMILES CC(C)(C)[Si](OS(=O)(=O)C(F)(F)F)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Brown
Quantity 1 g
UN Number 1760
Formula Weight 388.48
Percent Purity ≥98.0% (T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material tert-Butyldiphenylsilyl Trifluoromethanesulfonate
TSCA No
Recommended Storage Refrigerator