missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyldiphenylchlorosilane 97.0+%, TCI America™
Supplier: TCI America B122325ML
Specifications
| tert-Butyldiphenylchlorosilane | |
| Yellow | |
| C16H19ClSi | |
| 25 mL | |
| tert-butylchlorodiphenylsilane, tert-butyldiphenylchlorosilane, t-butyldiphenylchlorosilane, silane, chloro 1,1-dimethylethyl diphenyl, tert-butyl chloro diphenylsilane, t-butylchlorodiphenylsilane, tbdpscl, t-butyldiphenylsilyl chloride, tert-butyldiphenyl chlorosilane, unii-3beu48ui4e | |
| CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl | |
| 274.863 | |
| 274.86 | |
| Liquid |
| 58479-61-1 | |
| 125°C | |
| MFCD00000497 | |
| 2987 | |
| MHYGQXWCZAYSLJ-UHFFFAOYSA-N | |
| tert-butyl-chloro-diphenylsilane | |
| 94078 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 58479-61-1 | |
| 274.863 | |
| MHYGQXWCZAYSLJ-UHFFFAOYSA-N | |
| 94078 | |
| CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl |
| C16H19ClSi | |
| MFCD00000497 | |
| tert-butylchlorodiphenylsilane, tert-butyldiphenylchlorosilane, t-butyldiphenylchlorosilane, silane, chloro 1,1-dimethylethyl diphenyl, tert-butyl chloro diphenylsilane, t-butylchlorodiphenylsilane, tbdpscl, t-butyldiphenylsilyl chloride, tert-butyldiphenyl chlorosilane, unii-3beu48ui4e | |
| tert-butyl-chloro-diphenylsilane |
Safety and Handling
TSCA : Yes