missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldiphenylchlorosilane 97.0+%, TCI America™

Catalog No. p-7118931
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B12235ML 5 mL
B122325ML 25 mL
B1223100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B12235ML Supplier TCI America Supplier No. B12235ML
Only null left

Chemical Identifiers

CAS 58479-61-1
Molecular Formula C16H19ClSi
Molecular Weight (g/mol) 274.863
MDL Number MFCD00000497
InChI Key MHYGQXWCZAYSLJ-UHFFFAOYSA-N
Synonym tert-butylchlorodiphenylsilane, tert-butyldiphenylchlorosilane, t-butyldiphenylchlorosilane, silane, chloro 1,1-dimethylethyl diphenyl, tert-butyl chloro diphenylsilane, t-butylchlorodiphenylsilane, tbdpscl, t-butyldiphenylsilyl chloride, tert-butyldiphenyl chlorosilane, unii-3beu48ui4e
PubChem CID 94078
IUPAC Name tert-butyl-chloro-diphenylsilane
SMILES CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl

Specifications

Color Yellow
Boiling Point 125°C
Quantity 5 mL
UN Number 2987
Formula Weight 274.86
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyldiphenylchlorosilane