missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyldiphenylchlorosilane 97.0+%, TCI America™

Supplier:  TCI America B1223100ML

 View more versions of this product

Catalog No. B1223100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

tert-Butyldiphenylchlorosilane
Yellow
C16H19ClSi
100 mL
tert-butylchlorodiphenylsilane, tert-butyldiphenylchlorosilane, t-butyldiphenylchlorosilane, silane, chloro 1,1-dimethylethyl diphenyl, tert-butyl chloro diphenylsilane, t-butylchlorodiphenylsilane, tbdpscl, t-butyldiphenylsilyl chloride, tert-butyldiphenyl chlorosilane, unii-3beu48ui4e
CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl
274.863
274.86
Liquid
58479-61-1
125°C
MFCD00000497
2987
MHYGQXWCZAYSLJ-UHFFFAOYSA-N
tert-butyl-chloro-diphenylsilane
94078
≥97.0% (GC)

Chemical Identifiers

58479-61-1
274.863
MHYGQXWCZAYSLJ-UHFFFAOYSA-N
94078
CC(C)(C)[Si](C1=CC=CC=C1)(C2=CC=CC=C2)Cl
C16H19ClSi
MFCD00000497
tert-butylchlorodiphenylsilane, tert-butyldiphenylchlorosilane, t-butyldiphenylchlorosilane, silane, chloro 1,1-dimethylethyl diphenyl, tert-butyl chloro diphenylsilane, t-butylchlorodiphenylsilane, tbdpscl, t-butyldiphenylsilyl chloride, tert-butyldiphenyl chlorosilane, unii-3beu48ui4e
tert-butyl-chloro-diphenylsilane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions