missing translation for 'onlineSavingsMsg'
Learn More

(tert-Butyldimethylsilyl)acetylene 97.0+%, TCI America™

Catalog No. p-7123494
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
B43655G 5 g
B436525G 25 g
2 options

Catalog No. B43655G

Supplier: TCI America B43655G

Only null left

Chemical Identifiers

CAS 86318-61-8
Molecular Formula C8H16Si
Molecular Weight (g/mol) 140.301
MDL Number MFCD00191877
InChI Key RTYNRTUKJVYEIE-UHFFFAOYSA-N
Synonym tert-butyldimethylsilyl acetylene, tert-butyldimethylsilylacetylene, tert-butyl ethynyl dimethylsilane, ethynyl-t-butyldimethylsilane, tert-butyldimethylsilyl ethyne, tert-butyl-ethynyl-dimethyl-silane, silane, 1,1-dimethylethyl ethynyldimethyl, t-butyldimethylsilylacetylene, tert-butyldimethylsilylethyne, tert-butylethynyldimethylsilane
PubChem CID 2757281
IUPAC Name tert-butyl-ethynyl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)C#C

Specifications

Color Yellow
Boiling Point 110°C
Quantity 5 g
UN Number 1993
Formula Weight 140.30
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material (tert-Butyldimethylsilyl)acetylene