accessibility menu, dialog, popup

UserName

CH3CH3CH3SiCH3CH3HC

CAS RN 86318-61-8

IUPAC Name: tert-butyl(ethynyl)dimethylsilane
Synonyms: tert-butyl(ethynyl)dimethylsilane
Molecular Weight (g/mol): 140.3
Molecular Formula: C8H16Si
InChi Key: RTYNRTUKJVYEIE-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)C#C
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (4)