missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylbenzene 98.0+%, TCI America™

Catalog No. p-7118632
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B071525ML 25 mL
B0715500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B071525ML Supplier TCI America Supplier No. B071525ML
Only null left

Chemical Identifiers

CAS 98-06-6
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00008816
InChI Key YTZKOQUCBOVLHL-UHFFFAOYSA-N
Synonym t-butylbenzene, benzene, 1,1-dimethylethyl, benzene, tert-butyl, 2-methyl-2-phenylpropane, phenyltrimethylmethane, pseudobutylbenzene, dimethylethylbenzene, trimethylphenylmethane, 1,1-dimethylethylbenzene, tert-butyl benzene
PubChem CID 7366
IUPAC Name tert-butylbenzene
SMILES CC(C)(C)C1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 168°C
Quantity 25 mL
UN Number 2709
Formula Weight 134.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butylbenzene
EINECSNumber (3)-0011&(3)-0022
RTECSNumber CY9120000
TSCA Yes