missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylbenzene 98.0+%, TCI America™

Catalog No. B0715500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0715500ML 500 mL
B071525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0715500ML Supplier TCI America Supplier No. B0715500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-06-6
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00008816
InChI Key YTZKOQUCBOVLHL-UHFFFAOYSA-N
Synonym t-butylbenzene, benzene, 1,1-dimethylethyl, benzene, tert-butyl, 2-methyl-2-phenylpropane, phenyltrimethylmethane, pseudobutylbenzene, dimethylethylbenzene, trimethylphenylmethane, 1,1-dimethylethylbenzene, tert-butyl benzene
PubChem CID 7366
IUPAC Name tert-butylbenzene
SMILES CC(C)(C)C1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 168°C
Quantity 500 mL
UN Number 2709
Formula Weight 134.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butylbenzene
EINECSNumber (3)-0011&(3)-0022
RTECSNumber CY9120000
TSCA Yes