missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylbenzene, 99%

Catalog No. AC107870025
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107870025 2.5 L Glass bottle
AC107875000 500 mL Glass bottle
AC107870010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC107870025 Supplier Thermo Scientific Chemicals Supplier No. 107870025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 98-06-6 | C10H14 | 134.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-06-6
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
MDL Number MFCD00008816
InChI Key YTZKOQUCBOVLHL-UHFFFAOYSA-N
Synonym t-butylbenzene, benzene, 1,1-dimethylethyl, benzene, tert-butyl, 2-methyl-2-phenylpropane, phenyltrimethylmethane, pseudobutylbenzene, dimethylethylbenzene, trimethylphenylmethane, 1,1-dimethylethylbenzene, tert-butyl benzene
PubChem CID 7366
IUPAC Name tert-butylbenzene
SMILES CC(C)(C)C1=CC=CC=C1

Specifications

Melting Point -58.0°C
Color Colorless to Yellow
Density 0.8600g/mL
Boiling Point 169.0°C
Flash Point 44°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.492
Linear Formula C6H5C(CH3)3
Quantity 2.5 L
Beilstein 05, 415
Specific Gravity 0.86
Merck Index 15, 1553
Solubility Information Solubility in water: 0.03g/L water (20°C). Other solubilities: miscible with alcohol,ether and benzene
Viscosity 1.15 mPa.s (20°C)
Formula Weight 134.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material tert-Butylbenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-632-4

RUO – Research Use Only