missing translation for 'onlineSavingsMsg'
Learn More

tert-Butylbenzene, 99%

Catalog No. AAA12378AU
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA12378AU 1000 mL
AAA12378AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA12378AU Supplier Thermo Scientific Chemicals Supplier No. A12378AU
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 98-06-6 | C10H14 | 134.222 g/mol

tert-Butylbenzene is an useful intermediate for manufacturing agrochemicals and pharmaceuticals. tert-Butyl Toluene is used as a solvent for resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butylbenzene is an useful intermediate for manufacturing agrochemicals and pharmaceuticals. tert-Butyl Toluene is used as a solvent for resins.

Solubility
Insoluble in water(0.03g/100 mL at 20°C) .

Notes
Keep container tightly closed in a cool, dry and well-ventilated place. Store away from strong oxidizing agents, acids.

Chemical Identifiers

CAS 98-06-6
Molecular Formula C10H14
Molecular Weight (g/mol) 134.222
MDL Number MFCD00008816
InChI Key YTZKOQUCBOVLHL-UHFFFAOYSA-N
Synonym t-butylbenzene, benzene, 1,1-dimethylethyl, benzene, tert-butyl, 2-methyl-2-phenylpropane, phenyltrimethylmethane, pseudobutylbenzene, dimethylethylbenzene, trimethylphenylmethane, 1,1-dimethylethylbenzene, tert-butyl benzene
PubChem CID 7366
IUPAC Name tert-butylbenzene
SMILES CC(C)(C)C1=CC=CC=C1

Specifications

Melting Point -58°C
Density 0.867
Boiling Point 168°C to 169°C
Flash Point 34°C (93°F)
Odor Characteristic
Refractive Index 1.492
Quantity 1000 mL
UN Number UN2709
Beilstein 1421537
Merck Index 14,1551
Solubility Information Insoluble in water(0.03g/100 mL at 20°C) .
Formula Weight 134.22
Percent Purity 99%
Chemical Name or Material tert-Butylbenzene
Show More Show Less
Hazard Category H226-H315-H320
Hazard Statement GHS H Statement
H226-H332-H315-H319-H335
Flammable liquid and vapor.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: BUTYL BENZENES
EINECSNumber 202-632-4
RTECSNumber CY9120000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only