missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl vinylacetate, 97%

Catalog No. p-7037998
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5685903 1 g
AAH5685906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5685903 Supplier Thermo Scientific Chemicals Supplier No. H5685903
Only null left

CAS: 14036-55-6 | C8H14O2 | 142.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14036-55-6
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD06411714
InChI Key NGASWKRTXGWPNN-UHFFFAOYSA-N
Synonym tert-butyl 3-butenoate, tert-butyl vinylacetate, 3-butenoic acid tert-butyl ester, 3-butenoic acid, 1,1-dimethylethyl ester, t-butyl vinylacetate, tert-butyl3-butenoate, tert-butyl-3-butenoate, acmc-20ajl7, vinylacetic acid tert-butyl ester, but-3-enoic acid tert-butyl ester
PubChem CID 639778
IUPAC Name tert-butyl but-3-enoate
SMILES CC(C)(C)OC(=O)CC=C

Specifications

Density 0.877
Flash Point 31°C (88°F)
Refractive Index 1.413
Linear Formula CH2=CHCH2CO2C(CH3)3
Quantity 1 g
UN Number UN3272
Formula Weight 142.2
Percent Purity 97%
Chemical Name or Material tert-Butyl vinylacetate
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only