missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Sulfide 98.0+%, TCI America™

Catalog No. B0807500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0807500ML 500 mL
B080725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0807500ML Supplier TCI America Supplier No. B0807500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 107-47-1
Molecular Formula C8H18S
Molecular Weight (g/mol) 146.292
MDL Number MFCD00008839
InChI Key LNMBCRKRCIMQLW-UHFFFAOYSA-N
Synonym tert-butyl sulfide, di-tert-butyl sulfide, di-tert-butyl sulphide, propane, 2,2'-thiobis 2-methyl, t-butyl sulfide, 2,2,4,4-tetramethyl-3-thiapentane, di-t-butyl sulfide, 2-tert-butylsulfanyl-2-methylpropane, 2,2'-thiobis 2-methylpropane, di-tert-butyl polysulfide
PubChem CID 7872
IUPAC Name 2-tert-butylsulfanyl-2-methylpropane
SMILES CC(C)(C)SC(C)(C)C

Specifications

Color Colorless
Boiling Point 150°C
Quantity 500 mL
UN Number 1993
Formula Weight 146.29
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Sulfide
EINECSNumber (2)-0474
TSCA Yes