missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl propiolate, 98%

Catalog No. AAH5104803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5104803 1 g
AAH5104806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5104803 Supplier Thermo Scientific Chemicals Supplier No. H5104803

CAS: 13831-03-3 | C7H10O2 | 126.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13831-03-3
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD00060100
InChI Key XGTPDIIFEPTULX-UHFFFAOYSA-N
Synonym tert-butyl propiolate, propiolic acid tert-butyl ester, t-butyl propiolate, 2-propynoic acid, 1,1-dimethylethyl ester, boc-acetylene, tert-butyl propalate, tert-butyl propargylate, acmc-209chj, tert-butyl propiolate #
PubChem CID 543038
IUPAC Name tert-butyl prop-2-ynoate
SMILES CC(C)(C)OC(=O)C#C

Specifications

Melting Point 18°C to 20°C
Density 0.919
Boiling Point 52°C to 53°C (27 mmHg)
Flash Point 28°C (82°F)
Refractive Index 1.418
Linear Formula HC≡CCO2C(CH3)3
Quantity 1 g
UN Number UN3272
Beilstein 1747175
Sensitivity Air Sensitive
Formula Weight 126.16
Percent Purity 98%
Chemical Name or Material tert-Butyl propiolate
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only