missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Phenyl Carbonate 96.0+%, TCI America™

Catalog No. p-7123032
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35905G 5 g
B359025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35905G Supplier TCI America Supplier No. B35905G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6627-89-0
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00008804
InChI Key UXWVQHXKKOGTSY-UHFFFAOYSA-N
Synonym t-butyl phenyl carbonate, tert-butylphenylcarbonate, 2-methyl-2-propanyl phenyl carbonate, carbonic acid, 1,1-dimethylethyl phenyl ester, pubchem13300, acmc-209nuo, phenyl t-butyl carbonate, uxwvqhxkkogtsy-uhfffaoysa, tert-butyl phenyl carbonate, carbonic acid tert-butylphenyl ester
PubChem CID 81113
IUPAC Name tert-butyl phenyl carbonate
SMILES CC(C)(C)OC(=O)OC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 78°C
Quantity 5 g
Formula Weight 194.23
Percent Purity ≥96.0% (HPLC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Phenyl Carbonate