missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Phenyl Carbonate 96.0+%, TCI America™

Catalog No. b359025g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B359025G 25 g
B35905G 5 g
2 options

Catalog No. B359025G

Supplier: TCI America B359025G

Only null left

Chemical Identifiers

CAS 6627-89-0
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00008804
InChI Key UXWVQHXKKOGTSY-UHFFFAOYSA-N
Synonym t-butyl phenyl carbonate, tert-butylphenylcarbonate, 2-methyl-2-propanyl phenyl carbonate, carbonic acid, 1,1-dimethylethyl phenyl ester, pubchem13300, acmc-209nuo, phenyl t-butyl carbonate, uxwvqhxkkogtsy-uhfffaoysa, tert-butyl phenyl carbonate, carbonic acid tert-butylphenyl ester
PubChem CID 81113
IUPAC Name tert-butyl phenyl carbonate
SMILES CC(C)(C)OC(=O)OC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 78°C
Quantity 25 g
Formula Weight 194.23
Percent Purity ≥96.0% (HPLC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Phenyl Carbonate