missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl phenyl carbonate, 97%

CAS: 6627-89-0 | C11H14O3 | 194.23 g/mol

Supplier:  Thermo Scientific Chemicals H6086722

 View more versions of this product

Catalog No. AAH6086722


Only null left
Explore available promotions

Description

Description

tert-Butyl phenyl carbonate is used in determination of octanol-water partition coefficients by microemulsion electrokinetic chromatography. It is also used in the synthesis of 2-nitroindoles and as a reagent for mono-Boc protection of α,ω-diamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl phenyl carbonate is used in determination of octanol-water partition coefficients by microemulsion electrokinetic chromatography. It is also used in the synthesis of 2-nitroindoles and as a reagent for mono-Boc protection of α,ω-diamines.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents.
Specifications

Specifications

tert-Butyl phenyl carbonate
1.05
101°C (214°F)
(CH3)3COCOOC6H5
100 g
t-butyl phenyl carbonate, tert-butylphenylcarbonate, 2-methyl-2-propanyl phenyl carbonate, carbonic acid, 1,1-dimethylethyl phenyl ester, pubchem13300, acmc-209nuo, phenyl t-butyl carbonate, uxwvqhxkkogtsy-uhfffaoysa, tert-butyl phenyl carbonate, carbonic acid tert-butylphenyl ester
UXWVQHXKKOGTSY-UHFFFAOYSA-N
tert-butyl phenyl carbonate
81113
98%
6627-89-0
83°C
C11H14O3
MFCD00008804
2361602
Slightly soluble in water.
CC(C)(C)OC(=O)OC1=CC=CC=C1
194.23
194.23

Chemical Identifiers

6627-89-0
194.23
UXWVQHXKKOGTSY-UHFFFAOYSA-N
81113
CC(C)(C)OC(=O)OC1=CC=CC=C1
C11H14O3
MFCD00008804
t-butyl phenyl carbonate, tert-butylphenylcarbonate, 2-methyl-2-propanyl phenyl carbonate, carbonic acid, 1,1-dimethylethyl phenyl ester, pubchem13300, acmc-209nuo, phenyl t-butyl carbonate, uxwvqhxkkogtsy-uhfffaoysa, tert-butyl phenyl carbonate, carbonic acid tert-butylphenyl ester
tert-butyl phenyl carbonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 229-601-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only