missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl phenyl carbonate, 97%

Catalog No. AAH6086714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6086714 25 g
AAH6086722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6086714 Supplier Thermo Scientific Chemicals Supplier No. H6086714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6627-89-0 | C11H14O3 | 194.23 g/mol

tert-Butyl phenyl carbonate is used in determination of octanol-water partition coefficients by microemulsion electrokinetic chromatography. It is also used in the synthesis of 2-nitroindoles and as a reagent for mono-Boc protection of α,ω-diamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl phenyl carbonate is used in determination of octanol-water partition coefficients by microemulsion electrokinetic chromatography. It is also used in the synthesis of 2-nitroindoles and as a reagent for mono-Boc protection of α,ω-diamines.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 6627-89-0
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00008804
InChI Key UXWVQHXKKOGTSY-UHFFFAOYSA-N
Synonym t-butyl phenyl carbonate, tert-butylphenylcarbonate, 2-methyl-2-propanyl phenyl carbonate, carbonic acid, 1,1-dimethylethyl phenyl ester, pubchem13300, acmc-209nuo, phenyl t-butyl carbonate, uxwvqhxkkogtsy-uhfffaoysa, tert-butyl phenyl carbonate, carbonic acid tert-butylphenyl ester
PubChem CID 81113
IUPAC Name tert-butyl phenyl carbonate
SMILES CC(C)(C)OC(=O)OC1=CC=CC=C1

Specifications

Density 1.05
Boiling Point 83°C
Flash Point 101°C (214°F)
Linear Formula (CH3)3COCOOC6H5
Quantity 25 g
Beilstein 2361602
Solubility Information Slightly soluble in water.
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material tert-Butyl phenyl carbonate
EINECSNumber 229-601-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only