missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl peroxybenzoate, 98%

Catalog No. AAA1737336
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1737336 500 g
AAA1737314 25 g
AAA1737322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1737336 Supplier Thermo Scientific Chemicals Supplier No. A1737336

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 614-45-9 | C11H14O3 | 194.23 g/mol

tert-Butyl peroxybenzoate was employed as polymerization and cross-linking catalyst. It was also was employed as initiator during  grafting of 2,2,6,6-tetramethyl-1-piperidinyloxy (TEMPO)-4-oxyacetamido-(3 propyltriethoxysilane) to poly(ethylene co-octene and in preparation of conformal poly(cyclohexyl methacrylate) thin films via initiated chemical vapor deposition.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl peroxybenzoate was employed as polymerization and cross-linking catalyst. It was also was employed as initiator during  grafting of 2,2,6,6-tetramethyl-1-piperidinyloxy (TEMPO)-4-oxyacetamido-(3 propyltriethoxysilane) to poly(ethylene co-octene and in preparation of conformal poly(cyclohexyl methacrylate) thin films via initiated chemical vapor deposition.

Solubility
Soluble in ether, alcohol, ester, and ketones. Insoluble in water. 

Notes
 Stored and transported as a mixture with inert solids and as a solvent slurry.

Chemical Identifiers

CAS 614-45-9
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00008802
InChI Key GJBRNHKUVLOCEB-UHFFFAOYSA-N
Synonym tert-butyl peroxybenzoate, tert-butyl perbenzoate, t-butyl perbenzoate, chaloxyd tbpb, perbutyl z, esperox 10, novox, trigonox c, tert-butyl peroxy benzoate, terc.butylperbenzoan
PubChem CID 11966
IUPAC Name tert-butyl benzenecarboperoxoate
SMILES CC(C)(C)OOC(=O)C1=CC=CC=C1

Specifications

Melting Point 6°C to 8°C
Density 1.042
Boiling Point 75°C to 76°C (0.2 mmHg)
Flash Point 93°C (199°F)
Odor Sweet
Refractive Index 1.499
Quantity 500 g
UN Number UN3103
Beilstein 1342734
Solubility Information Soluble in ether,alcohol,ester,and ketones. Insoluble in water.
Formula Weight 194.23
Percent Purity 98%
Chemical Name or Material tert-Butyl peroxybenzoate
Show More Show Less
Hazard Category H227-H315-H317-H319-H332-H335
Hazard Statement GHS H Statement
H200-H242-H302-H315-H319-H227
Unstable explosives.
Heating may cause a fire.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.2; Packing Group: II; Proper Shipping Name: ORGANIC PEROXIDE TYPE C, LIQUID
EINECSNumber 210-382-2
RTECSNumber SD9450000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only