missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl Nitrite 90.0+%, TCI America™

Catalog No. p-7133769
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N035725ML 25 mL
N0357250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N035725ML Supplier TCI America Supplier No. N035725ML
Only null left

Chemical Identifiers

CAS 540-80-7
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00002055
InChI Key IOGXOCVLYRDXLW-UHFFFAOYSA-N
Synonym t-butyl nitrite, 1,1-dimethylethyl nitrite, tert-butylnitrite, nitrous acid, 1,1-dimethylethyl ester, unii-4780h7u8lu, ccris 2098, alpha,alpha-dimethylethyl nitrite, nitrous acid, t-butyl ester, 2-methyl-2-propylnitrit, ch3 3cono
PubChem CID 10906
IUPAC Name tert-butyl nitrite
SMILES CC(C)(C)ON=O

Specifications

Color Yellow
Boiling Point 63°C
Quantity 25 mL
UN Number 2351
Formula Weight 103.12
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material tert-Butyl Nitrite
RTECSNumber RA0802000
TSCA No
Recommended Storage Refrigerator