accessibility menu, dialog, popup

UserName

CAS RN 540-80-7

IUPAC Name: tert-butyl nitrite
Synonyms: 2-methyl-2-propylnitrit
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: IOGXOCVLYRDXLW-UHFFFAOYSA-N
SMILES: CC(C)(C)ON=O
Boiling Point 
  • (2)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (4)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (5)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)