missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-Allylcarbamate 98.0+%, TCI America™

Supplier:  TCI America B361825G

Quantity 25 g

 View more versions of this product

Catalog No. B361825G


Only null left

Description

Description

Specifications

Chemical Identifiers

78888-18-3
157.213
AWARHXCROCWEAK-UHFFFAOYSA-N
4067274
CC(C)(C)OC(=O)NCC=C
C8H15NO2
MFCD00191870
tert-butyl allylcarbamate, tert-butyl n-allylcarbamate, tert-butyl n-prop-2-en-1-yl carbamate, n-boc-allylamine, t-butyl n-allylcarbamate, carbamic acid, 2-propenyl-, 1,1-dimethylethyl ester, acmc-209pfu, allyl tert-butyl carbamate, tert-butyl-n-allylcarbamate, n-tertbutoxycarbonyl allyl amine
tert-butyl N-prop-2-enylcarbamate

Specifications

39°C
78888-18-3
White-Yellow
MFCD00191870
AWARHXCROCWEAK-UHFFFAOYSA-N
tert-butyl N-prop-2-enylcarbamate
157.213
157.21
Crystalline Powder
tert-Butyl N-Allylcarbamate
49°C
C8H15NO2
tert-butyl allylcarbamate, tert-butyl n-allylcarbamate, tert-butyl n-prop-2-en-1-yl carbamate, n-boc-allylamine, t-butyl n-allylcarbamate, carbamic acid, 2-propenyl-, 1,1-dimethylethyl ester, acmc-209pfu, allyl tert-butyl carbamate, tert-butyl-n-allylcarbamate, n-tertbutoxycarbonyl allyl amine
CC(C)(C)OC(=O)NCC=C
25 g
4067274
≥98.0% (GC)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions