missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-Allylcarbamate 98.0+%, TCI America™

Catalog No. p-7123051
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36185G 5 g
B361825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36185G Supplier TCI America Supplier No. B36185G

Chemical Identifiers

CAS 78888-18-3
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD00191870
InChI Key AWARHXCROCWEAK-UHFFFAOYSA-N
Synonym tert-butyl allylcarbamate, tert-butyl n-allylcarbamate, tert-butyl n-prop-2-en-1-yl carbamate, n-boc-allylamine, t-butyl n-allylcarbamate, carbamic acid, 2-propenyl-, 1,1-dimethylethyl ester, acmc-209pfu, allyl tert-butyl carbamate, tert-butyl-n-allylcarbamate, n-tertbutoxycarbonyl allyl amine
PubChem CID 4067274
IUPAC Name tert-butyl N-prop-2-enylcarbamate
SMILES CC(C)(C)OC(=O)NCC=C

Specifications

Melting Point 39°C
Boiling Point 49°C
Color White-Yellow
Quantity 5 g
Formula Weight 157.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material tert-Butyl N-Allylcarbamate