missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-[4-(aminomethyl)phenyl]carbamate, 97%, Thermo Scientific™

Catalog No. mo01028da
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MO01028DA 1 g
MO01028EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MO01028DA Supplier Thermo Scientific Chemicals Supplier No. MO01028DA

Chemical Identifiers

CAS 220298-96-4
Molecular Formula C12H18N2O2
Molecular Weight (g/mol) 222.29
MDL Number MFCD02183573
InChI Key URXUHALBOWYXJZ-UHFFFAOYSA-N
Synonym tert-butyl n-4-aminomethyl phenyl carbamate, 4-aminomethyl-1-n-boc-aniline, tert-butyl 4-aminomethyl phenyl carbamate, 4-boc-amino benzylamine, 4-aminomethy-1-n-boc-aniline, 4-n-boc-amino benzylamine, 4-tert-butoxycarbonylamino benzylamine, 4-aminomethyl-n-boc-aniline, tert-butyl-n-4-aminomethyl phenyl carbamate
PubChem CID 2794659
IUPAC Name tert-butyl N-[4-(aminomethyl)phenyl]carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=C(CN)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 222.29
Percent Purity 97%
Chemical Name or Material tert-Butyl N-[4-(aminomethyl)phenyl]carbamate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.