missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl N-(3-thienyl)carbamate, 97%, Thermo Scientific™

Catalog No. CC00092DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC00092DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC00092DA Supplier Thermo Scientific Chemicals Supplier No. CC00092DA
Only null left

Chemical Identifiers

CAS 19228-91-2
Molecular Formula C9H13NO2S
Molecular Weight (g/mol) 199.268
MDL Number MFCD01928808
InChI Key PRWYQCYSADTIBZ-UHFFFAOYSA-N
Synonym tert-butyl thiophen-3-ylcarbamate, tert-butyl n-3-thienyl carbamate, tert-butyl 3-thienylcarbamate, tert-butyl n-thiophen-3-yl carbamate, tert-butoxy-n-3-thienyl carboxamide, carbamic acid,n-3-thienyl-, 1,1-dimethylethyl ester, n-boc-3-aminothiophene, tert-butyl thien-3-ylcarbamate, 3-aminothiophene, n-boc protected
PubChem CID 736476
IUPAC Name tert-butyl N-thiophen-3-ylcarbamate
SMILES CC(C)(C)OC(=O)NC1=CSC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 199.27
Percent Purity 97%
Chemical Name or Material Tert-butyl N-(3-thienyl)carbamate